Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCEAL8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | TCEAL8 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
TCEAL8 Polyclonal specifically detects TCEAL8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TCEAL8 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
90843 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
MGC45400, TCEA-like protein 8, transcription elongation factor A (SII)-like 8, transcription elongation factor A protein-like 8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human TCEAL8 (NP_699164.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title