Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCEANC2 Mouse anti-Human, Clone: OTI2F12, lyophilized, TrueMAB™
SDP

Catalog No. 501669449
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-9449 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-9449 Supplier OriGene Supplier No. CF809365
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

TCEANC2 (Transcription Elongation Factor A N-Terminal And Central Domain Containing 2) is a Protein Coding gene.

Specifications

Antigen TCEANC2
Applications Western Blot
Classification Monoclonal
Clone OTI2F12
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TCEANC2
Gene Accession No. Q96MN5
Gene Alias C1orf83
Gene Symbols TCEANC2
Host Species Mouse
Immunogen Full length human recombit protein of human TCEANC2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127428
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.