Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEB1 Recombinant Rabbit Monoclonal Antibody (HL1683)
GREENER_CHOICE

Catalog No. PIMA547140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547140 Supplier Invitrogen™ Supplier No. MA547140
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1683
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ELOC
Gene Accession No. P83940, Q15369
Gene Alias 2610043E24Rik; 2610301I15Rik; AA407206; AI987979; AW049146; eloC; elongation factor SIII p15 subunit; elongin 15 kDa subunit; elongin C; elongin-C; RNA polymerase II transcription factor SIII subunit C; SIII; SIII p15; Stromal membrane-associated protein SMAP1B homolog; Tceb1; transcription elongation factor B (SIII), polypeptide 1; transcription elongation factor B (SIII), polypeptide 1 (15kDa, elongin C); transcription elongation factor B polypeptide 1; transcription elongation factor B polypeptide 1; elongin-C; transcription elongation factor B subunit 1
Gene Symbols ELOC
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human TCEB1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100628309, 477914, 67923, 6921
Target Species Human, Mouse, Canine, Feline
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.