Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCEB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550802
Description
The antibody detects endogenous levels of total TCEB1 protein.
TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation.
Specifications
TCEB1 | |
Polyclonal | |
Unconjugated | |
ELOC | |
2610043E24Rik; 2610301I15Rik; AA407206; AI987979; AW049146; eloC; elongation factor SIII p15 subunit; elongin 15 kDa subunit; elongin C; elongin-C; RNA polymerase II transcription factor SIII subunit C; SIII; SIII p15; Stromal membrane-associated protein SMAP1B homolog; Tceb1; transcription elongation factor B (SIII), polypeptide 1; transcription elongation factor B (SIII), polypeptide 1 (15kDa, elongin C); transcription elongation factor B polypeptide 1; transcription elongation factor B polypeptide 1; elongin-C; transcription elongation factor B subunit 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64525, 67923, 6921 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P83940, P83941, Q15369 | |
ELOC | |
Full length fusion protein of Human TCEB1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction