Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110465 Supplier Invitrogen™ Supplier No. PA5110465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDVFLMIRRH KTTIFTDAKE SSTVFELKRI VEGILKRPPD EQRLYKDDQL LDDGKTLGEC GFTSQTARPQ APATVGLAFR ADDTFEALCI EPFSSPPELP DVMKPQDSGS SANEQAVQ.

SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEB2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ELOB
Gene Accession No. P62869, Q15370
Gene Alias 0610040H15Rik; 18 kDa, elongin B; cb162; ELOB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; elongin-B; hypothetical protein LOC617277; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII; SIII p18; TCEB 2; Tceb2; tceb2 protein; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2; transcription elongation factor B (SIII), polypeptide 2 (18 kDa, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); transcription elongation factor B polypeptide 2; transcription elongation factor B polypeptide 2; elongin-B; transcription elongation factor B subunit 2; wu:fb47e08
Gene Symbols ELOB
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67673, 6923
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.