Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCEB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548686
Description
TCEB2 Polyclonal Antibody for Western Blot
SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).
Specifications
TCEB2 | |
Polyclonal | |
Unconjugated | |
ELOB | |
0610040H15Rik; 18 kDa, elongin B; cb162; ELOB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; elongin-B; hypothetical protein LOC617277; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII; SIII p18; TCEB 2; Tceb2; tceb2 protein; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2; transcription elongation factor B (SIII), polypeptide 2 (18 kDa, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); transcription elongation factor B polypeptide 2; transcription elongation factor B polypeptide 2; elongin-B; transcription elongation factor B subunit 2; wu:fb47e08 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
6923 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q15370 | |
ELOB | |
KLH conjugated synthetic peptide between 21-48 amino acids from the Central region of human TCEB2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction