Learn More
Invitrogen™ TCEB2 Polyclonal Antibody

Description
Immunogen sequence: MDVFLMIRRH KTTIFTDAKE SSTVFELKRI VEGILKRPPD EQRLYKDDQL LDDGKTLGEC GFTSQTARPQ APATVGLAFR ADDTFEALCI EPFSSPPELP DVMKPQDSGS SANEQAVQ; Positive Samples: 22Rv1, NCI-H460, MCF7; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | TCEB2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ELOB |
| Gene Accession No. | Q15370 |
| Gene Alias | 0610040H15Rik; 18 kDa, elongin B; cb162; ELOB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; elongin-B; hypothetical protein LOC617277; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII; SIII p18; TCEB 2; Tceb2; tceb2 protein; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2; transcription elongation factor B (SIII), polypeptide 2 (18 kDa, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); transcription elongation factor B polypeptide 2; transcription elongation factor B polypeptide 2; elongin-B; transcription elongation factor B subunit 2; wu:fb47e08 |
| Gene Symbols | ELOB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.