Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCEB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 107791AP150UL

Description
SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).Specifications
TCEB2 | |
Polyclonal | |
Unconjugated | |
ELOB | |
ELOB, Elongin 18 kDa subunit, Elongin B, SIII, SIII p18, TCEB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67673, 6923, 81807 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62869, P62870, Q15370 | |
Elob | |
TCEB2 Fusion Protein Ag1228 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction