Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCERG1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300214UV
Description
TCERG1 Polyclonal antibody specifically detects TCERG1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TCERG1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CA150TATA box-binding protein-associated factor 2S, co-activator of 150 kDa, TAF2SMGC133200, TATA box binding protein (TBP)-associated factor, RNA polymerase II, S, 150kD, TATA box-binding protein-associated factor 2S, transcription elongation regulator 1, Transcription factor CA150, Urn1 | |
A synthetic peptide made to a portion of human transcription elongation regulator 1 (CA150) encoded within exon 1 (LocusLink ID 10915). | |
0.1 mL | |
Cellular Markers, Transcription Factors and Regulators | |
10915 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction