Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCERG1L Antibody (05), Alexa Fluor™ 488 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305852AF488
Description
TCERG1L Monoclonal antibody specifically detects TCERG1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TCERG1L | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 5730476P14Rik, FLJ38950, MGC126584, transcription elongation regulator 1-like, transcription elongation regulator 1-like protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the C-terminus of the Human TCERG1L. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 05 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 256536 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction