Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF-12/HTF4 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002338AF488
Description
TCF-12/HTF4 Polyclonal antibody specifically detects TCF-12/HTF4 in Human samples. It is validated for Western BlotSpecifications
TCF-12/HTF4 | |
Polyclonal | |
Western Blot | |
BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4TCF-12, transcription factor 12, Transcription factor HTF-4 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Transcription Factor 12 using the numbering given in entry NP_996919.1 (GeneID 6938). | |
0.1 mL | |
Immunology | |
6938 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction