Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF-12/HTF4 Antibody (OTI4D6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274473F
Description
TCF-12/HTF4 Monoclonal specifically detects TCF-12/HTF4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
TCF-12/HTF4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TCF12 | |
Recombinant protein fragment corresponding to amino acids 1-250 of human TCF-12/HTF4 (NP_996919) produced in E.coli. | |
0.1 mL | |
6938.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4D6 | |
Immunohistochemistry | |
BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4TCF-12, transcription factor 12, Transcription factor HTF-4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction