Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF-2/HNF-1 beta Antibody (SR2073), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | TCF-2/HNF-1 beta |
---|---|
Clone | SR2073 |
Dilution | Western Blot 1:1000-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
TCF-2/HNF-1 beta Monoclonal antibody specifically detects TCF-2/HNF-1 beta in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TCF-2/HNF-1 beta | |
Western Blot 1:1000-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FJHN, hepatocyte nuclear factor 1-beta, HNF1 beta A, HNF1 homeobox B, HNF-1B, HNF1beta, HNF-1-beta, HNF2, Homeoprotein LFB3, LFB3, LF-B3, MODY5HPC11, TCF-2, TCF2transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor, Transcription factor 2, transcription factor 2, hepatic, Variant hepatic nuclear factor 1, vHNF1 | |
A synthesized peptide derived from human TCF-2/HNF-1 beta (Uniprot #: P35680) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2073 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Diabetes Research | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6928 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title