Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF-2/HNF-1 beta Antibody (SR2149), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321992100UL
Description
TCF-2/HNF-1 beta Monoclonal antibody specifically detects TCF-2/HNF-1 beta in Human samples. It is validated for Western BlotSpecifications
TCF-2/HNF-1 beta | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR2149 | |
Western Blot 1:500-1:2000 | |
FJHN, hepatocyte nuclear factor 1-beta, HNF1 beta A, HNF1 homeobox B, HNF-1B, HNF1beta, HNF-1-beta, HNF2, Homeoprotein LFB3, LFB3, LF-B3, MODY5HPC11, TCF-2, TCF2transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor, Transcription factor 2, transcription factor 2, hepatic, Variant hepatic nuclear factor 1, vHNF1 | |
A synthesized peptide derived from human TCF-2/HNF-1 beta (Uniprot #: P35680) | |
100 μg | |
Diabetes Research | |
6928 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction