Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF-3/4 Mouse anti-Mouse, Human, Clone: 6F12, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $463.74
Specifications
Antigen | TCF-3/4 |
---|---|
Clone | 6F12 |
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE199325
|
MilliporeSigma
MABE199325UL |
25 μL |
Each for $182.31
|
|
|||||
MABE1993100
|
MilliporeSigma
MABE1993100UL |
100 μL |
Each for $463.74
|
|
|||||
Description
TCF-3/4 Monoclonal antibody specifically detects TCF-3/4 in Mouse, Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.Specifications
TCF-3/4 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
6F12 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human | |
His-tagged recombinant fragment corresponding to 301 amino acids from the N-terminal half of human TCF-4. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title