Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF-7/TCF-1 Mouse anti-Mouse, Human, Alexa Fluor 488, Clone: S33-966, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567018
This item is not returnable.
View return policy
Description
TCF-7/TCF-1 Monoclonal antibody specifically detects TCF-7/TCF-1 in Mouse, Human samples. It is validated for Intracellular staining (Flow Cytometry)
Specifications
TCF-7/TCF-1 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Flow Cytometry | |
S33-966 | |
Alexa Fluor 488 | |
TCF7; Transcription factor 7; TCF-1; TCF1; T-cell factor 1 | |
Mouse TCF-7 Recombinant Protein | |
RUO | |
Mouse, Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction