Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCF12 Monoclonal Antibody (OTI2E11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527198
Description
TCF12 Monoclonal Antibody for Western Blot, IHC (P)
TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.
Specifications
TCF12 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99081 | |
TCF12 | |
Full length human recombinant protein of TCF12 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2E11 | |
Unconjugated | |
TCF12 | |
A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6938 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction