Learn More
TCF12 Mouse anti-Human, Clone: OTI3C11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809697

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.Specifications
TCF12 | |
Monoclonal | |
Unconjugated | |
TCF12 | |
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6938 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3C11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q99081 | |
TCF12 | |
Full length human recombit protein of human TCF12 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.