Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578193 Supplier Invitrogen™ Supplier No. PA578193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Jurkat, Jurkat nuclear extract Predicted Reactivity: Mouse (89%), Rat (88%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF12
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene TCF12
Gene Accession No. Q99081
Gene Alias A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4
Gene Symbols TCF12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TCF12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.