Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCF15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566313 Supplier Invitrogen™ Supplier No. PA566313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLSNQRKGGGR RDLGGSCLKV RGVAPLRGPR R Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

TCF15 is found in the nucleus and may be involved in the early transcriptional regulation of patterning of the mesoderm. The basic helix-loop-helix protein requires dimerization with another basic helix-loop-helix protein for efficient DNA binding.The protein encoded by this gene is found in the nucleus and may be involved in the early transcriptional regulation of patterning of the mesoderm. The encoded basic helix-loop-helix protein requires dimerization with another basic helix-loop-helix protein for efficient DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF15
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCF15
Gene Accession No. Q12870
Gene Alias basic helix-loop-helix transcription factor 15; bHLHa40; Bhlhec2; bHLH-EC2; Class A basic helix-loop-helix protein 40; EC2; LOW QUALITY PROTEIN: transcription factor 15; Meso1; par1; par1 protein; paraxis; Protein bHLH-EC2; TCF15; TCF-15; Transcription factor 15; transcription factor 15 (basic helix-loop-helix)
Gene Symbols TCF15
Host Species Rabbit
Immunogen Recombinant Human TCF15. Recombinant protein control fragment (Product #RP-103088).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.