Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF19 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$160.07 - $406.62

Specifications

Antigen TCF19
Concentration 0.5 mg/mL
Dilution Immunohistochemistry-Paraffin 5-10 ug/ml
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224586SS
SDP
View Documents
Novus Biologicals
NBP224586SS
0.025 mg
Each for $160.07
Only null left
Add to Cart
 
NBP224586
SDP
View Documents
Novus Biologicals
NBP224586
0.1 mg
Each for $406.62
Only null left
Add to Cart
 
Description

Description

TCF19 Polyclonal specifically detects TCF19 in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TCF19
Immunohistochemistry-Paraffin 5-10 ug/ml
Polyclonal
Rabbit
Core ESC Like Genes, Stem Cell Markers
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
SC1(TCF19)-6, SC1-1, SC1SC1(TCF19)-7, SCI(TCF19)-4, TCF-19, transcription factor 19, Transcription factor SC1
TCF19
IgG
Affinity Purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in beta I and beta II Tubulin.
0.5 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Primate
Q9Y242
6941
A synthetic peptide corresponding to amino acids 259-287 of human TCF19 was used as immunogen for the antibody, GenBank no Q9Y242.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.