Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCF19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104188 Supplier Invitrogen™ Supplier No. PA5104188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TCF19.

TCF-19 (transcription factor 19), also known as SC1 or SC1-1, is a 345 amino acid protein that contains one FHA domain, a proline-rich domain and one PHD-type zinc finger. Localizing to the nucleus, TCF-19 is a growth regulated protein that is believed to function as a trans-activating factor with a role in the transcription of genes involved in the late stages of cell cycle progression (G1 to S transition or entry of cells into G2 and mitosis). TCF-19 is expressed preferentially in the G1-S phase of the cell cycle. The gene encoding TCF-19 localizes to a critical region on chromosome 6 that has been associated with psoriasis vulgaris, a disorder of the skin that is characterized by hyperproliferation of epidermal cells. This suggests that TCF-19, via its regulation of late cell cycle-specific genes, may play a role in the development of this disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF19
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TCF19
Gene Accession No. Q99KJ5, Q9Y242
Gene Alias 5730403J10Rik; AW495861; DAMA-213L4.5; SC 1; SC1; SC1 protein homolog; SC1(TCF19)-6; SC1(TCF19)-7; SC1-1; SCI(TCF19)-4; TCF 19; TCF19; TCF-19; Transcription factor 19; transcription factor 19-like protein; Transcription factor SC1
Gene Symbols TCF19
Host Species Rabbit
Immunogen A synthesized peptide derived from human TCF19(Accession Q9Y242), corresponding to amino acid residues P241-P291.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106795, 406195, 6941
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.