Learn More
Invitrogen™ TCF19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110468
Description
Immunogen sequence: MLPCFQLLRI GGGRGGDLYT FHPPAGAGCT YRLGHRADLC DVALRPQQEP GLISGIHAEL HAEPRGDDWR VSLEDHSSQG TLVNNVRLPR GHRLELSDGD LLTFGPEGPP GTSPSEFYFM FQQVRVKPQD FAAITIPRSR GEARVGAGFR PMLPSQGAPQ RPLSTFSPAP KATLILNSIG SLSKLRPQPL TFSPSWGGPK SLPVPAPPGE MGTTPSAPPQ RNRRKSVHRV LAELDDESEP PENPPPVLME PRKKLRVDKA PLTPTGNRRG RPRKYPVSAP MAPPAVGGGE.
TCF-19 (transcription factor 19), also known as SC1 or SC1-1, is a 345 amino acid protein that contains one FHA domain, a proline-rich domain and one PHD-type zinc finger. Localizing to the nucleus, TCF-19 is a growth regulated protein that is believed to function as a trans-activating factor with a role in the transcription of genes involved in the late stages of cell cycle progression (G1 to S transition or entry of cells into G2 and mitosis). TCF-19 is expressed preferentially in the G1-S phase of the cell cycle. The gene encoding TCF-19 localizes to a critical region on chromosome 6 that has been associated with psoriasis vulgaris, a disorder of the skin that is characterized by hyperproliferation of epidermal cells. This suggests that TCF-19, via its regulation of late cell cycle-specific genes, may play a role in the development of this disorder.
Specifications
| TCF19 | |
| Polyclonal | |
| Unconjugated | |
| TCF19 | |
| 5730403J10Rik; AW495861; DAMA-213L4.5; SC 1; SC1; SC1 protein homolog; SC1(TCF19)-6; SC1(TCF19)-7; SC1-1; SCI(TCF19)-4; TCF 19; TCF19; TCF-19; Transcription factor 19; transcription factor 19-like protein; Transcription factor SC1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 106795, 406195, 6941 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99KJ5, Q9Y242 | |
| TCF19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human TCF19 (NP_009040.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.