Learn More
Invitrogen™ TCF2 Monoclonal Antibody (CL0374)

Description
Immunogen sequence: KEVLVQALEE LLPSPNFGVK LETLPLSPGS GAEPDTKPVF HTLTNGHAKG RLSGDEGSED GDDYDTPPIL KELQALNTEE AAEQRAEVDR MLSEDPWRAA KMIKGYMQQH Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%. Binds to an epitope located within the peptide sequence TPPILKELQA as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | TCF2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL0374 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HNF1B |
| Gene Accession No. | P35680 |
| Gene Alias | FJHN; Hepatocyte nuclear factor 1-beta; hepatocyte nuclear factor-1 beta; HNF1 beta A; HNF1 homeobox B; HNF1B; Hnf-1b; Hnf1beta; HNF-1Beta; HNF-1-beta; HNF2; homeoprotein LFB3; HPC11; LFB3; LF-B3; MODY5; Tcf2; Tcf-2; Transcription factor 2; Transcription factor 2 hepatic; Transcription factor 2 hepatic; LF-B3; variant hepatic nuclear factor; transcription factor 2, hepatic; Transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor; variant hepatic nuclear factor 1; vHNF1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.