Learn More
Invitrogen™ TCF2 Polyclonal Antibody

Description
The antibody detects endogenous level of total HNF1B protein.
Specifications
Specifications
| Antigen | TCF2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | HNF1B |
| Gene Accession No. | P23899, P27889, P35680 |
| Gene Alias | FJHN; Hepatocyte nuclear factor 1-beta; hepatocyte nuclear factor-1 beta; HNF1 beta A; HNF1 homeobox B; HNF1B; Hnf-1b; Hnf1beta; HNF-1Beta; HNF-1-beta; HNF2; homeoprotein LFB3; HPC11; LFB3; LF-B3; MODY5; Tcf2; Tcf-2; Transcription factor 2; Transcription factor 2 hepatic; Transcription factor 2 hepatic; LF-B3; variant hepatic nuclear factor; transcription factor 2, hepatic; Transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor; variant hepatic nuclear factor 1; vHNF1 |
| Gene Symbols | HNF1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.