Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF21 Polyclonal Antibody, Huabio

Catalog No. 502544584
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4584 Supplier HUABIO RESEARCH INC Supplier No. HA500188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TCF21 Polyclonal antibody specifically detects TCF21 in Human samples. It is validated for Western Blot

Specifications

Antigen TCF21
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TCF21
Gene Accession No. O43680
Gene Alias BHLHA23; capsulin; class A basic helix-loop-helix protein 23; epc; Epicardin; hypothetical protein LOC558148; Msf-1; MyoRa2; Pod1; pod-1; podocyte-expressed 1; TCF21; TCF-21; Transcription factor 21; zgc:123313
Host Species Rabbit
Immunogen Synthetic peptide within human TCF21 aa 125-175.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.