Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCF3 Recombinant Rabbit Monoclonal Antibody (HL1954)
GREENER_CHOICE

Catalog No. PIMA547233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547233 Supplier Invitrogen™ Supplier No. MA547233
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

TCF3 (E2A) is a transcription factor that plays major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Heterodimers between E2A and tissue-specific basic helix-loop-helix (bHLH) Dimers bind DNA on E-box motifs: 5'- CANNTG-3'. Binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer. Deletions in E2A have been observed in a subset of pre-B-cell acute lymphoblastic leukemia (B-ALL) cases. Two alternatively spliced human isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1954
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TCF3
Gene Accession No. P15923
Gene Alias A1; AA408400; AGM8; Alf2; AW209082; bHLHb21; class B basic helix-loop-helix protein 21; E12; E12/E47; E2A; E47; helix-loop-helix protein HE47; immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; ITF1; Kappa-E2-binding factor; Me2; MGC129647; MGC129648; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; Pan; Pan1; Pan2; pancreas specific transcription factor 1c; Ptf1c; TCF3; TCF-3; Tcfe2a; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; transcription factor A1; transcription factor E2a; Transcription factor E2-alpha; transcription factor ITF-1; Transcription regulator Pan; VDIR; VDR interacting repressor; vitamin D receptor-interacting repressor
Gene Symbols TCF3
Host Species Rabbit
Immunogen Recombinant fragment of human TCF3 / E2A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6929
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.