Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCF3/TCF4 Monoclonal Antibody (6F12-3)

Catalog No. PIMA125418
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125418 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125418 Supplier Invitrogen™ Supplier No. MA125418

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TCF 4 (T cell factor 4) is a member of the TCF/LEF (lymphoid enhancer factor) family of transcription factors. Members of this family along with b catenin, are involved in mediating Wnt signaling by transactivating downstream genes. TCF 4 generates transcriptionally active complexes with b catenin in colon carcinoma cells. Expression of TCF 4 is highest in the CNS and intestinal epithelium where Wnt signaling also plays an essential role in development. Aberrant TCF 4 transcription activity has been implicated in carcinogenesis of intestinal and mammary epithlia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF3/TCF4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6F12-3
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation TBS with 30% glycerol and 0.08% sodium azide; pH 7.4
Gene Tcf4
Gene Accession No. P15806, P15884, P15923, Q60722
Gene Alias 5730422P05Rik; A1; AA408400; AGM8; ALF2; ASP-I2; AW209082; bHLHb19; BHLHB21; Class A helix-loop-helix transcription factor ME2; class B basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 21; E12; E12/E47; E2.2; E2-2; E2A; E47; FECD3; helix-loop-helix protein HE47; immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF1; ITF-1; ITF2; ITF-2; ITF-2b; kappa-E2-binding factor; Me2; MITF-2; MITF-2A; MITF-2B; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; Pan1; Pan2; PTHS; SEF2; SEF-2; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; SL3-3 enhancer factor 2; Tcf3; TCF-3; TCF4; TCF-4; Tcfe2a; TFE; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R; transcription factor A1; transcription factor E2a; Transcription factor E2-alpha; transcription factor ITF-1; VDIR; VDR interacting repressor; vitamin D receptor-interacting repressor
Gene Symbols TCF3, Tcf4
Host Species Mouse
Immunogen His-tagged fusion protein corresponding to amino acids 31-331 of human TCF.
Purification Method Protein G
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21413, 21423, 6925, 6929
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.