Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF4 Mouse anti-Human, Clone: TCF4/1705, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | TCF4 |
---|---|
Clone | TCF4/1705 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Protein Array |
Applications | Western Blot, ELISA, Peptide Array |
Description
TCF4 Monoclonal specifically detects TCF4 in Human samples. It is validated for Western Blot, ELISA, Protein Array.Specifications
TCF4 | |
0.2 mg/ml | |
Western Blot, ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
6925 | |
IgG2a κ | |
Protein A or G purified |
TCF4/1705 | |
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 | |
Recombinant human TCF4 protein fragment (around aa 365-671) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title