Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF4 Mouse anti-Human, Clone: TCF4/2252, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | TCF4 |
---|---|
Clone | TCF4/2252 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
TCF4 | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 | |
Recombinant human TCF4 protein fragment (around aa 365-671) (exact sequence is proprietary) (Uniprot: P15884 ) | |
Primary | |
Store at 4°C. |
TCF4/2252 | |
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P15884 | |
6925 | |
IgG2c κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title