Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCF4 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142391
Description
Additional Information: This antibody is expected to recognise the reported isoforms a and b (NP_001077431.1; NP_003190.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
TCF4 is transcription factor 4, a basic helix-turn-helix transcription factor. The encoded protein recognizes an Ephrussi-box ('E-box') binding site ('CANNTG')-a motif first identified in immunoglobulin enhancers. This gene is expressed predominantly in pre-B-cells, although it is found in other tissues as well. Defects in this gene are a cause of Pitt-Hopkins syndrome.
Specifications
| TCF4 | |
| Polyclonal | |
| Unconjugated | |
| TCF4 | |
| 5730422P05Rik; ASP-I2; bHLHb19; class A helix-loop-helix transcription factor ME2; Class B basic helix-loop-helix protein 19; E2 2; E2.2; E2-2; FECD3; Immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF 2; ITF2; ITF-2; ITF-2b; ME2; MGC149723; MGC149724; MITF-2; MITF-2A; MITF-2B; PTHS; R8f DNA-binding protein; RITF-2; SEF 2; SEF2; SEF-2; SEF2 1; SEF2 1A; SEF2 1B; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; SL3-3 enhancer factor 2; TCF 4; TCF4; TCF-4; TFE; Transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 6925 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P15884 | |
| TCF4 | |
| Peptide with sequence C-NGGHPSPSRNYGDGT (aa 61-75). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction