Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525659
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525659 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525659 Supplier Invitrogen™ Supplier No. PA525659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

TCF4 is transcription factor 4, a basic helix-turn-helix transcription factor. The encoded protein recognizes an Ephrussi-box ('E-box') binding site ('CANNTG')-a motif first identified in immunoglobulin enhancers. This gene is expressed predominantly in pre-B-cells, although it is found in other tissues as well. Defects in this gene are a cause of Pitt-Hopkins syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF4
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TCF4
Gene Accession No. P15884, Q60722
Gene Alias 5730422P05Rik; ASP-I2; bHLHb19; class A helix-loop-helix transcription factor ME2; Class B basic helix-loop-helix protein 19; E2 2; E2.2; E2-2; FECD3; Immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF 2; ITF2; ITF-2; ITF-2b; ME2; MGC149723; MGC149724; MITF-2; MITF-2A; MITF-2B; PTHS; R8f DNA-binding protein; RITF-2; SEF 2; SEF2; SEF-2; SEF2 1; SEF2 1A; SEF2 1B; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; SL3-3 enhancer factor 2; TCF 4; TCF4; TCF-4; TFE; Transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R
Gene Symbols TCF4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 48-73 amino acids from the N-terminal region of human TCF4.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21413, 6925
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.