Learn More
Invitrogen™ TCF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588125
Description
Immunogen sequence: MHHQQRMAAL GTDKELSDLL DFSAMFSPPV SSGKNGPTSL ASGHFTGSNV EDRSSSGSWG NGGHPSPSRN YGDGTPYDHM TSRDLGSHDN LSPPFVNSRI QSKTERGSYS SYGRESNLQG CHQQSLLGGD MDMGNPGTLS PTKPGSQYYQ YSSNNPRRRP LHSSAMEVQT KKVRKVPPGL PSSVYAPSAS TADYNRDSPG YPSSKPATST Positive Samples: A375, K-562; Cellular Location: Nucleus.
TCF4 is transcription factor 4, a basic helix-turn-helix transcription factor. The encoded protein recognizes an Ephrussi-box ('E-box') binding site ('CANNTG')-a motif first identified in immunoglobulin enhancers. This gene is expressed predominantly in pre-B-cells, although it is found in other tissues as well. Defects in this gene are a cause of Pitt-Hopkins syndrome.
Specifications
TCF4 | |
Polyclonal | |
Unconjugated | |
TCF4 | |
5730422P05Rik; ASP-I2; bHLHb19; class A helix-loop-helix transcription factor ME2; Class B basic helix-loop-helix protein 19; E2 2; E2.2; E2-2; FECD3; Immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF 2; ITF2; ITF-2; ITF-2b; ME2; MGC149723; MGC149724; MITF-2; MITF-2A; MITF-2B; PTHS; R8f DNA-binding protein; RITF-2; SEF 2; SEF2; SEF-2; SEF2 1; SEF2 1A; SEF2 1B; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; SL3-3 enhancer factor 2; TCF 4; TCF4; TCF-4; TFE; Transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R | |
Rabbit | |
Affinity Chromatography | |
RUO | |
21413, 6925, 84382 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.95 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P15884, Q60722, Q62655 | |
TCF4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human TCF4 (NP_0031901). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.