Learn More
TCF4 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | TCF4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | TCF4 |
| Gene Accession No. | P15884 |
| Gene Alias | 5730422P05Rik; ASP-I2; BHLHB19; Class A helix-loop-helix transcription factor ME2; Class B basic helix-loop-helix protein 19; E2 2; E2.2; E2-2; FECD3; Immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF 2; Itf2; ITF-2; ITF-2b; ME2; MGC149723; MGC149724; MITF-2; MITF-2A; MITF-2B; PTHS; R8f DNA-binding protein; RITF-2; SEF 2; SEF2; SEF-2; SEF2 1; SEF2 1A; SEF2 1B; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; SL3-3 enhancer factor 2; TCF 4; Tcf4; TCF-4; TFE; Transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R |
| Gene Symbols | TCF4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.