Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCF4/TCF12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536151
Description
This antibody detects endogenous protein at a molecular weight of 60 and 72 kDa. Purity is >95% by SDS-PAGE.
T cell factors (TCFs) comprise a family of DNA-binding transcriptional activators that are essential for lymphoid cell development. These transcription factors are activated by the Wnt-1 and Wingless pathways and are characterized by the presence of a conserved protein motif, the high mobility group(HMG) 1 box, which mediates DNA binding. TCF-4 mainly localizes in the cytoplasm and is transported into the nucleus directly bound to beta-catenin in a cooperative manner. This TCF-4/ beta-catenin complex induces expression of Wnt target genes, including multiple cancer-associated genes.
Specifications
TCF4/TCF12 | |
Polyclonal | |
Unconjugated | |
TCF4 | |
5730422P05Rik; A130037E08Rik; Alf1; ASP-I2; bHLHb19; bHLHb20; Class A helix-loop-helix transcription factor ME1; class A helix-loop-helix transcription factor ME2; Class B basic helix-loop-helix protein 19; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E2.2; E2-2; E-box-binding protein; FECD3; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; Immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF2; ITF-2; ITF-2b; ME1; ME2; Meb; MITF-2; MITF-2A; MITF-2B; PTHS; R8f DNA-binding protein; REB; RITF-2; salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; SEF2; SEF-2; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; SL3-3 enhancer factor 2; TCF12; TCF-12; Tcf4; TCF-4; TFE; transcription factor 12; Transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R; transcription factor HTF-4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21406, 21413, 25720, 6925, 6938, 84382 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P15884, P51514, Q60722, Q61286, Q62655, Q99081 | |
TCF12, TCF4 | |
Synthetic peptide corresponding to amino acids 580-630 of human TCF-4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction