Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102691 Supplier Thermo Scientific Supplier No. PA5102691
Add to Cart
Add to Cart

TCF7 Polyclonal Antibody

TCF7 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TCF7
Gene Accession No. P36402, Q00417
Gene Alias AI465550; HMG-box; OTTHUMP00000159390; OTTHUMP00000159391; T cell factor-1; T-cell factor 1; T-cell specific; T-cell-specific transcription factor 1; Tcf1; TCF-1; Tcf7; TCF-7; Transcription factor 7; transcription factor 7 (T-cell specific, HMG-box); transcription factor 7, T cell specific; transcription factor 7, T-cell specific
Gene Symbols TCF7
Host Species Rabbit
Immunogen A synthesized peptide derived from human TCF7(Accession P36402), corresponding to amino acid residues S6-V56.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21414, 363595, 6932
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.