Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591923 Supplier Invitrogen™ Supplier No. PA591923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse spleen; Cellular Location: Nucleus Immunogen sequence: ELKSSLVNES EGAAGGAGIP GVPGAGAGAR GEAEALGREH AAQRLFPDKL PEPLEDGLKA PECTSGMYKE TVYSAFNLLM HYPPPSGAGQ HPQPQPPLHK A.

TCF7 (transcriptional factor 7) is a transcriptional activator involved in T-cell lymphocyte differentiation. Necessary for the survival of CD4+ and CD8+ immature thymocytes. Isoforms for TCF7 lacking the N-terminal CTNNB1 binding domain cannot fulfill this role. TCF7 binds to the T-lymphocyte-specific enhancer element (5'-WWCAAAG-3') found in the promoter of the CD3E gene. It may also act as feedback transcriptional repressor of CTNNB1 and TCF7L2 target genes. TLE1, TLE2, TLE3, and TLE4 repress transactivation mediated by TCF7 and CTNNB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TCF7
Gene Accession No. P36402, Q00417
Gene Alias AI465550; HMG-box; OTTHUMP00000159390; OTTHUMP00000159391; T cell factor-1; T-cell factor 1; T-cell specific; T-cell-specific transcription factor 1; Tcf1; TCF-1; Tcf7; TCF-7; Transcription factor 7; transcription factor 7 (T-cell specific, HMG-box); transcription factor 7, T cell specific; transcription factor 7, T-cell specific
Gene Symbols TCF7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human TCF1/TCF7 (NP_0031932).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21414, 363595, 6932
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.