Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF7 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993518
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3518 10 μL
50-199-3517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3518 Supplier Active Motif Supplier No. 61580
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TCF7 (transcriptional factor 7) is a transcriptional activator involved in T-cell lymphocyte differentiation. Necessary for the survival of CD4+ and CD8+ immature thymocytes. Isoforms for TCF7 lacking the N-terminal CTNNB1 binding domain cannot fulfill this role. TCF7 binds to the T-lymphocyte-specific enhancer element (5'-WWCAAAG-3') found in the promoter of the CD3E gene. It may also act as feedback transcriptional repressor of CTNNB1 and TCF7L2 target genes. TLE1, TLE2, TLE3, and TLE4 repress transactivation mediated by TCF7 and CTNNB1.

Specifications

Antigen TCF7
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene TCF7
Gene Accession No. P36402
Gene Alias AI465550; HMG-box; OTTHUMP00000159390; OTTHUMP00000159391; T cell factor-1; T-cell factor 1; T-cell specific; T-cell-specific transcription factor 1; Tcf1; Tcf-1; Tcf7; TCF-7; Transcription factor 7; transcription factor 7 (T-cell specific, HMG-box); transcription factor 7, T cell specific; transcription factor 7, T-cell specific
Gene Symbols TCF7
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the N-terminal region of human TCF7.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6932
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.