Learn More
Invitrogen™ TCF7 Recombinant Rabbit Monoclonal Antibody (4K0X6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555567

Description
Immunogen Sequence: ELKSSLVNES EGAAGGAGIP GVPGAGAGAR GEAEALGREH AAQRLFPDKL PEPLEDGLKA PECTSGMYKE TVYSAFNLLM HYPPPSGAGQ HPQPQPPLHK A.
TCF7 (transcriptional factor 7) is a transcriptional activator involved in T-cell lymphocyte differentiation. Necessary for the survival of CD4+ and CD8+ immature thymocytes. Isoforms for TCF7 lacking the N-terminal CTNNB1 binding domain cannot fulfill this role. TCF7 binds to the T-lymphocyte-specific enhancer element (5'-WWCAAAG-3') found in the promoter of the CD3E gene. It may also act as feedback transcriptional repressor of CTNNB1 and TCF7L2 target genes. TLE1, TLE2, TLE3, and TLE4 repress transactivation mediated by TCF7 and CTNNB1.
Specifications
TCF7 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P36402, Q00417 | |
TCF7 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human TCF1/TCF7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
4K0X6 | |
Unconjugated | |
TCF7 | |
AI465550; HMG-box; OTTHUMP00000159390; OTTHUMP00000159391; T cell factor-1; T-cell factor 1; T-cell specific; T-cell-specific transcription factor 1; Tcf1; TCF-1; Tcf7; TCF-7; Transcription factor 7; transcription factor 7 (T-cell specific, HMG-box); transcription factor 7, T cell specific; transcription factor 7, T-cell specific | |
Rabbit | |
Protein A | |
RUO | |
21414, 363595, 6932 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.