Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF7/TCF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25821325UL
Description
TCF7/TCF1 Polyclonal specifically detects TCF7/TCF1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TCF7/TCF1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
FLJ36364, MGC47735, T-cell factor 1, T-cell specific, TCF1, TCF-7, transcription factor 7 (T-cell specific, HMG-box) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
TCF7 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KKCIRYLPGEGRCPSPVPSDDSALGCPGSPAPQDSPSYHLLPRFPTELLTSPAE | |
25 μL | |
Signal Transduction, Wnt Signaling Pathway | |
6932 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction