Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF7/TCF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | TCF7/TCF1 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TCF7/TCF1 Polyclonal specifically detects TCF7/TCF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TCF7/TCF1 | |
Polyclonal | |
Rabbit | |
Signal Transduction, Wnt Signaling Pathway | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
6932 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KQVHRPLQTPDLSGFYSLTSGSMGQLPHTVSWFTHPSLMLGSGVPGHPAAIPHPAIVPPSGKQELQPFDRNLKTQAESKAEKEAK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
FLJ36364, MGC47735, T-cell factor 1, T-cell specific, TCF1, TCF-7, transcription factor 7 (T-cell specific, HMG-box) | |
TCF7 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title