Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF7L1/TCF3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298907AF350
Description
TCF7L1/TCF3 Polyclonal specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western Blot.Specifications
TCF7L1/TCF3 | |
Polyclonal | |
Western Blot | |
HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box) | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human TCF7L1/TCF3. | |
0.1 mL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway | |
83439 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction