Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF7L1/TCF3 Antibody (SR2086), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321655100UL
Description
TCF7L1/TCF3 Monoclonal antibody specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western BlotSpecifications
TCF7L1/TCF3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR2086 | |
Western Blot 1:500-1:2000 | |
HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box) | |
A synthesized peptide derived from human TCF7L1/TCF3 (Uniprot #: Q9HCS4) | |
100 μg | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway | |
83439 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction