Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF7L2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119083H
Description
TCF7L2 Polyclonal specifically detects TCF7L2 in Human samples. It is validated for Western Blot.Specifications
TCF7L2 | |
Polyclonal | |
Western Blot | |
Q9NQB0 | |
TCF7L2 | |
Synthetic peptide made to an internal portion of human TCF7L2 (within residues 150-200). [Swiss-Prot# Q9NQB0] | |
Affinity Purified | |
RUO | |
Primary | |
Human. Immunogen sequence has 93% homology to mouse and bovine. | |
Store at 4C in the dark. |
Western Blot | |
HRP | |
PBS with No Preservative | |
HMG box transcription factor 4, hTCF-4, T-cell factor 4, T-cell factor-4 variant A, T-cell factor-4 variant B, T-cell factor-4 variant C, T-cell factor-4 variant D, T-cell factor-4 variant E, T-cell factor-4 variant H, T-cell factor-4 variant I, T-cell factor-4 variant J, T-cell factor-4 variant K, T-cell factor-4 variant L, T-cell factor-4 variant M, T-cell factor-4 variant X2, T-cell-specific transcription factor 4, TCF-4T-cell factor-4 variant F, TCF4T-cell factor-4 variant G, transcription factor 7-like 2, transcription factor 7-like 2 (T-cell specific, HMG-box) | |
Rabbit | |
55 kDa | |
0.1 mL | |
Signal Transduction, Wnt Signaling Pathway | |
6934 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction