Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCF9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567476
Description
Immunogen sequence: ESEPDDHEKRI PFTLRPQTLR QRMAEESISR NEETSEESQE DEKQDTWEQQ QMRKAVKIIE ERDIDLSCGS GSSKVKKFDT SISFP Highest antigen sequence identity to the following orthologs - mouse 72%, rat 73%.
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
Specifications
TCF9 | |
Polyclonal | |
Unconjugated | |
GCFC2 | |
A130099G21; AW146020; C2orf3; DNABF; GC binding factor; GC bindng factor; GCF; GCF2; GCFC2; GC-rich sequence DNA binding factor 2; GC-rich sequence DNA-binding factor; GC-rich sequence DNA-binding factor 2; RGD1304792; TCF9; TCF-9; Transcription factor 9; transcription factor 9 (binds GC-rich sequences) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6936 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P16383 | |
GCFC2 | |
Recombinant Human TCF9. Recombinant protein control fragment (Product #RP-108129). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction