Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCFL5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24917425UL
Description
TCFL5 Polyclonal antibody specifically detects TCFL5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
TCFL5 | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
bHLHe82, CHA, Cha transcription factor, CHAMGC46135, E2BP1, E2BP-1HPV-16 E2 binding protein 1, Figlb, HPV-16 E2-binding protein 1, transcription factor-like 5 (basic helix-loop-helix), transcription factor-like 5 protein | |
This antibody was developed against a recombinant protein corresponding to amino acids: KRNRSRMRQLDTNVERRALGEIQNVGEGATATQGAWQSSESSQANLGEQAQSGPQGGRSQRRERHNRMERDRRRRIRI | |
25 μL | |
Cell Cycle and Replication | |
10732 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction