Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCFL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584246 Supplier Invitrogen™ Supplier No. PA584246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRNRSRMRQL DTNVERRALG EIQNVGEGAT ATQGAWQSSE SSQANLGEQA QSGPQGGRSQ RRERHNRMER DRRRRIRI Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 88%.

TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCFL5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCFL5
Gene Accession No. Q9UL49
Gene Alias AV260129; bHLHe82; CHA; Cha transcription factor; E2BP1; E2BP-1; factor in the germline beta; FIG beta; Figlb; HPV-16 E2-binding protein 1; LOW QUALITY PROTEIN: transcription factor-like 5 protein; MGC46135; RGD1305899; SOSF1; TCFL5; transcription factor like 5; transcription factor-like 5 (basic helix-loop-helix); transcription factor-like 5 protein
Gene Symbols TCFL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCFL5. Recombinant protein control fragment (Product #RP-101724).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.