Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCHH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120392 Supplier Invitrogen™ Supplier No. PA5120392
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, Jurakt, HeLa, NIH/3T3, Mouse brain, Mouse kidnay, Mouse lung, Mouse pancreas. Immunogen sequence: RARCDGKESL LQDRRQEEDQ RRFEPRDRQL EEEPGQRRRQ KRQEQERELA EGEEQSEKQE RLEQRDRQRR DEELWRQRQE WQEREERRAE EEQLQSCKGH ETEEFPDEEQ LRRRELLELR RKGREEKQQQ RRERQDRVFQ EEEEKEWRKR ETVLRKEEEK LQEEEPQRQR ELQEEEEQLR KLERQELRRE RQEEEQQQQR L.

This protein is an intermediate filament-associated protein that associates in regular arrays with keratin intermediate filaments (KIF) of the inner root sheath cells of the hair follicle and the granular layer of the epidermis. It later becomes cross-linked to KIF by isodipeptide bonds. It may serve as scaffold protein, together with involucrin, in the organization of the cell envelope or even anchor the cell envelope to the KIF network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCHH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TCHH
Gene Accession No. Q07283
Gene Alias AHF; AI597080; hair follicle protein AHF; Tchh; THH; THL; TRHY; Trichohyalin
Gene Symbols TCHH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-300 of human TCHH (NP_009044.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7062, 99681
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.