Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCHH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | TCHH |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121558
|
Invitrogen™
PA5121558 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5121557
|
Invitrogen™
PA5121557 |
100 μL |
Each for $511.50
|
|
|||||
Description
This protein is an intermediate filament-associated protein that associates in regular arrays with keratin intermediate filaments (KIF) of the inner root sheath cells of the hair follicle and the granular layer of the epidermis. It later becomes cross-linked to KIF by isodipeptide bonds. It may serve as scaffold protein, together with involucrin, in the organization of the cell envelope or even anchor the cell envelope to the KIF network.Specifications
TCHH | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
Q07283 | |
310588, 7062 | |
Phe1851-Pro1943 (Accession Q07283), with N-terminal His Tag. | |
Primary | |
-20°C | |
TCHH |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
AHF; AI597080; hair follicle protein AHF; Tchh; THH; THL; TRHY; Trichohyalin | |
TCHH | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title