Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCHHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584597 Supplier Invitrogen™ Supplier No. PA584597
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDQEPCSVER GAVYSSPLYQ YLQEKILQQT NVTQEEHQKQ VQIAQASGPE LCSVSLTSEI SDCSVFFNYS QASQPYTRGL PLDESPAGAQ ETPAPQ Highest antigen sequence indentity to the following orthologs: Mouse - 25%, Rat - 25%.

This gene belongs to the S100 fused-type protein (SFTP) gene family, and is located in a cluster of SFTP genes on chromosome 1q21. Several members of this family have been implicated in the development of complex skin disorders. This gene is evolutionarily conserved; its expression appears to be hair-specific and spatially restricted within the distal inner root sheath of the hair follicle. It thus may have an important role in hair morphogenesis. [provided by RefSeq, Aug 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCHHL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCHHL1
Gene Accession No. Q5QJ38
Gene Alias basalin; intermediate filament-associated protein; Protein S100-A17; S100 calcium binding protein A17; S100 calcium-binding protein A17; S100A17; TCHHL1; THHL1; trichohyalin like 1; trichohyalin-like 1; trichohyalin-like protein 1
Gene Symbols TCHHL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCHHL1. Recombinant protein control fragment (Product #RP-107239).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.